[(3-Methyl-1H-1,2,4-triazol-5-yl)thio]acetic acid
Supplier: Matrix Scientific
037115-500MG
101842-710EA
75.38
USD
101842-710
[(3-Methyl-1H-1,2,4-triazol-5-yl)thio]acetic acid
[(3-Methyl-1H-1,2,4-triazol-5-yl)thio]acetic acid
Hazard: Irritant
Formula:
C₅H₇N₃O₂S |
MDL Number:
MFCD03536501 CAS Number: 64679-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...