5-Methyl-1-(3-methylbenzyl)-1H-pyrazol-3-amine

Supplier: Matrix Scientific

037114-500MG
101842-708EA 289.8 USD
101842-708
5-Methyl-1-(3-methylbenzyl)-1H-pyrazol-3-amine
5-Methyl-1-(3-methylbenzyl)-1H-pyrazol-3-amine

Hazard: Irritant

Formula: C₁₂H₁₅N₃
MDL Number: MFCD04971118
CAS Number: 956729-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR