1-Benzyl-1H-pyrazol-3-amine

Supplier: Matrix Scientific

037113-500MG
101842-706EA 289.8 USD
101842-706
1-Benzyl-1H-pyrazol-3-amine
1-Benzyl-1H-pyrazol-3-amine

Hazard: Irritant

Formula: C₁₀H₁₁N₃
MDL Number: MFCD03422517
CAS Number: 21377-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR