2-(3,4-Dihydroisoquinolin-2(1H)-yl)ethanamine

Supplier: Matrix Scientific

037110-500MG
101842-698EA 196.92 USD
101842-698
2-(3,4-Dihydroisoquinolin-2(1H)-yl)ethanamine
2-(3,4-Dihydroisoquinolin-2(1H)-yl)ethanamine

Hazard: Irritant

Formula: C₁₁H₁₆N₂
MDL Number: MFCD01717314
CAS Number: 53356-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR