4-(Piperazin-1-ylsulfonyl)aniline

Supplier: Matrix Scientific

037098-500MG
101842-670EA 290.77 USD
101842-670
4-(Piperazin-1-ylsulfonyl)aniline
4-(Piperazin-1-ylsulfonyl)aniline

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₂S
MDL Number: MFCD03444527
CAS Number: 69249-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR