Methyl-3-hydroxypiperidine-1-carboxylate

Supplier: Matrix Scientific

037097-500MG
101842-668EA 461.54 USD
101842-668
Methyl-3-hydroxypiperidine-1-carboxylate
Methyl-3-hydroxypiperidine-1-carboxylate

Hazard: Irritant

Formula: C₇H₁₃NO₃
MDL Number: MFCD11053970
CAS Number: 80613-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR