Tetrahydro-2H-pyran-3-carboxamide

Supplier: Matrix Scientific

037096-500MG
101842-666EA 336.95 USD
101842-666
Tetrahydro-2H-pyran-3-carboxamide
Tetrahydro-2H-pyran-3-carboxamide

Hazard: Irritant

Formula: C₆H₁₁NO₂
MDL Number: MFCD12027052
CAS Number: 18729-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR