3-(2-Hydroxy-1-naphthyl)propanoic acid

Supplier: Matrix Scientific

037093-500MG
101842-658EA 290.77 USD
101842-658
3-(2-Hydroxy-1-naphthyl)propanoic acid
3-(2-Hydroxy-1-naphthyl)propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₂O₃
MDL Number: MFCD08164671
CAS Number: 10441-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR