1-(3-Aminophenyl)-5-methylpyrrolidin-2-one

Supplier: Matrix Scientific

037092-500MG
101842-656EA 375.38 USD
101842-656
1-(3-Aminophenyl)-5-methylpyrrolidin-2-one
1-(3-Aminophenyl)-5-methylpyrrolidin-2-one

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MDL Number: MFCD10686558
CAS Number: 1033693-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR