2-(2-Oxotetrahydrofuran-3-yl)-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

037086-500MG
101842-642EA 217.35 USD
101842-642
2-(2-Oxotetrahydrofuran-3-yl)-1H-isoindole-1,3(2H)-dione
2-(2-Oxotetrahydrofuran-3-yl)-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₂H₉NO₄
MDL Number: MFCD00276776
CAS Number: 42473-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR