4-(4-Amino-2,5-dimethylbenzyl)-2,5-dimethylaniline

Supplier: Matrix Scientific

037082-500MG
101842-634EA 217.35 USD
101842-634
4-(4-Amino-2,5-dimethylbenzyl)-2,5-dimethylaniline
4-(4-Amino-2,5-dimethylbenzyl)-2,5-dimethylaniline

Hazard: Irritant

Formula: C₁₇H₂₂N₂
MDL Number: MFCD06622277
CAS Number: 5339-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR