Tetrahydro-4H-pyran-4-one oxime

Supplier: Matrix Scientific

Synonyms: Dihydro-2H-pyran-4(3H)-one oxime

037077-500MG
101842-624EA 193.85 USD
101842-624
Tetrahydro-4H-pyran-4-one oxime
Tetrahydro-4H-pyran-4-one oxime

Hazard: Irritant

Formula: C₅H₉NO₂
MW: 115.13 g/mol
MDL Number: MFCD09998842
CAS Number: 61128-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR