Diisobutyl-2,3-dihydroxysuccinate

Supplier: Matrix Scientific

037075-500MG
101842-620EA 109.25 USD
101842-620
Diisobutyl-2,3-dihydroxysuccinate
Diisobutyl-2,3-dihydroxysuccinate

Hazard: Irritant

Formula: C₁₂H₂₂O₆
MDL Number: MFCD00129104
CAS Number: 4054-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR