5,5-Dimethylpyrrolidin-2-one

Supplier: Matrix Scientific

037074-500MG
101842-618EA 83.08 USD
101842-618
5,5-Dimethylpyrrolidin-2-one
5,5-Dimethylpyrrolidin-2-one

Hazard: Irritant

Formula: C₆H₁₁NO
MDL Number: MFCD00128876
CAS Number: 5165-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR