(1E)-1-(2,4-Dihydroxyphenyl)ethanone oxime

Supplier: Matrix Scientific

037073-500MG
101842-616EA 193.85 USD
101842-616
(1E)-1-(2,4-Dihydroxyphenyl)ethanone oxime
(1E)-1-(2,4-Dihydroxyphenyl)ethanone oxime

Hazard: Irritant

Formula: C₈H₉NO₃
MDL Number: MFCD00043493
CAS Number: 6134-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR