3-(3,5-Dimethyl-1H-pyrazol-1-yl)-2-methylpropanenitrile

Supplier: Matrix Scientific

037067-500MG
101842-604EA 193.85 USD
101842-604
3-(3,5-Dimethyl-1H-pyrazol-1-yl)-2-methylpropanenitrile
3-(3,5-Dimethyl-1H-pyrazol-1-yl)-2-methylpropanenitrile

Hazard: Irritant

Formula: C₉H₁₃N₃
MDL Number: MFCD06739385
CAS Number: 1006319-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR