2,2'-Dichloro-1,1'-biphenyl-4,4'-diamine

Supplier: Matrix Scientific

037065-500MG
101842-600EA 221.54 USD
101842-600
2,2'-Dichloro-1,1'-biphenyl-4,4'-diamine
2,2'-Dichloro-1,1'-biphenyl-4,4'-diamine

Hazard: Irritant

Formula: C₁₂H₁₀Cl₂N₂
MDL Number: MFCD00209459
CAS Number: 84-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR