1-Methyl-4-nitro-1H-pyrazole-3,5-dicarboxylic acid

Supplier: Matrix Scientific

037064-500MG
101842-598EA 289.8 USD
101842-598
1-Methyl-4-nitro-1H-pyrazole-3,5-dicarboxylic acid
1-Methyl-4-nitro-1H-pyrazole-3,5-dicarboxylic acid

Hazard: Irritant

Formula: C₆H₅N₃O₆
MDL Number: MFCD03424117
CAS Number: 159427-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR