3-Hydroxy-1,8-naphthalic anhydride

Supplier: Matrix Scientific

037062-500MG
101842-594EA 106.15 USD
101842-594
3-Hydroxy-1,8-naphthalic anhydride
3-Hydroxy-1,8-naphthalic anhydride

Hazard: Irritant

Formula: C₁₂H₆O₄
MW: 214.17 g/mol
MDL Number: MFCD00059759
CAS Number: 23204-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR