1,2-Bis(1H-benzimidazol-2-yl)ethane-1,2-diol

Supplier: Matrix Scientific

037060-500MG
101842-590EA 217.35 USD
101842-590
1,2-Bis(1H-benzimidazol-2-yl)ethane-1,2-diol
1,2-Bis(1H-benzimidazol-2-yl)ethane-1,2-diol

Hazard: Irritant

Formula: C₁₆H₁₄N₄O₂
MDL Number: MFCD00128421
CAS Number: 3314-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR