(5-Thien-2-yl-1H-pyrazol-4-yl)methanol

Supplier: Matrix Scientific

037058-500MG
101842-586EA 116.92 USD
101842-586
(5-Thien-2-yl-1H-pyrazol-4-yl)methanol
(5-Thien-2-yl-1H-pyrazol-4-yl)methanol

Hazard: Irritant

Formula: C₈H₈N₂OS
MDL Number: MFCD09864606
CAS Number: 194546-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR