N-Hydroxy-4-methoxy-N-phenylbenzamide

Supplier: Matrix Scientific

037056-500MG
101842-582EA 144.9 USD
101842-582
N-Hydroxy-4-methoxy-N-phenylbenzamide
N-Hydroxy-4-methoxy-N-phenylbenzamide

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
MDL Number: MFCD00134775
CAS Number: 13664-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR