3-Pyridin-2-ylpropanenitrile

Supplier: Matrix Scientific

037055-500MG
101842-580EA 650.77 USD
101842-580
3-Pyridin-2-ylpropanenitrile
3-Pyridin-2-ylpropanenitrile

Hazard: Irritant

Formula: C₈H₈N₂
MDL Number: MFCD06656817
CAS Number: 35549-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR