1-(4-Nitrophenyl)-1H-1,2,3-triazole-4,5-dicarbohydrazide

Supplier: Matrix Scientific

036023-500MG
101840-538EA 289.8 USD
101840-538
1-(4-Nitrophenyl)-1H-1,2,3-triazole-4,5-dicarbohydrazide
1-(4-Nitrophenyl)-1H-1,2,3-triazole-4,5-dicarbohydrazide

Hazard: Irritant

Formula: C₁₀H₁₀N₈O₄
MDL Number: MFCD01433961
CAS Number: 156361-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR