2-[(2,6-Dibromo-4-methylphenyl)amino]acetohydrazide

Supplier: Matrix Scientific

036022-500MG
101840-536EA 290.77 USD
101840-536
2-[(2,6-Dibromo-4-methylphenyl)amino]acetohydrazide
2-[(2,6-Dibromo-4-methylphenyl)amino]acetohydrazide

Hazard: Irritant

Formula: C₉H₁₁Br₂N₃O
MW: 337.03 g/mol
MDL Number: MFCD00448181
CAS Number: 882760-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR