2-[(2-Methylphenyl)amino]propanohydrazide

Supplier: Matrix Scientific

036020-500MG
101840-532EA 290.77 USD
101840-532
2-[(2-Methylphenyl)amino]propanohydrazide
2-[(2-Methylphenyl)amino]propanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₅N₃O
MDL Number: MFCD00465010
CAS Number: 1396972-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR