4-(3,5-Dimethyl-1H-pyrazol-1-yl)-6-hydrazino-N-(4-methylphenyl)-1,3,5-triazin-2-amine

Supplier: Matrix Scientific

036019-500MG
101840-530EA 289.8 USD
101840-530
4-(3,5-Dimethyl-1H-pyrazol-1-yl)-6-hydrazino-N-(4-methylphenyl)-1,3,5-triazin-2-amine
4-(3,5-Dimethyl-1H-pyrazol-1-yl)-6-hydrazino-N-(4-methylphenyl)-1,3,5-triazin-2-amine

Hazard: Irritant

Formula: C₁₅H₁₈N₈
MDL Number: MFCD02255692
CAS Number: 1158287-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR