2-[(2-Bromophenyl)amino]propanohydrazide

Supplier: Matrix Scientific

036018-500MG
101840-528EA 217.35 USD
101840-528
2-[(2-Bromophenyl)amino]propanohydrazide
2-[(2-Bromophenyl)amino]propanohydrazide

Hazard: Irritant

Formula: C₉H₁₂BrN₃O
MDL Number: MFCD04002118
CAS Number: 1396994-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR