2-[(4-Iodophenyl)amino]propanohydrazide

Supplier: Matrix Scientific

036017-500MG
101840-526EA 290.77 USD
101840-526
2-[(4-Iodophenyl)amino]propanohydrazide
2-[(4-Iodophenyl)amino]propanohydrazide

Hazard: Irritant

Formula: C₉H₁₂IN₃O
MDL Number: MFCD02255640
CAS Number: 1397002-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR