5-(tert-Butyl)-2-(chloromethyl)-1,3-benzoxazole

Supplier: Matrix Scientific

036012-500MG
101840-516EA 217.35 USD
101840-516
5-(tert-Butyl)-2-(chloromethyl)-1,3-benzoxazole
5-(tert-Butyl)-2-(chloromethyl)-1,3-benzoxazole

Hazard: Irritant

Formula: C₁₂H₁₄ClNO
MDL Number: MFCD07366538
CAS Number: 65999-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR