2-(Chloromethyl)-1,3-benzoxazole-5-sulfonamide

Supplier: Matrix Scientific

036011-500MG
101840-514EA 290.77 USD
101840-514
2-(Chloromethyl)-1,3-benzoxazole-5-sulfonamide
2-(Chloromethyl)-1,3-benzoxazole-5-sulfonamide

Hazard: Irritant

Formula: C₈H₇ClN₂O₃S
MDL Number: MFCD09261474
CAS Number: 936074-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR