5-Chloro-2-(chloromethyl)-1,3-benzoxazole

Supplier: Matrix Scientific

036008-500MG
101840-508EA 101.2 USD
101840-508
5-Chloro-2-(chloromethyl)-1,3-benzoxazole
5-Chloro-2-(chloromethyl)-1,3-benzoxazole

Hazard: Irritant

Formula: C₈H₅Cl₂NO
MDL Number: MFCD05663796
CAS Number: 63842-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR