3-Methoxypyridin-2-ol

Supplier: Matrix Scientific

036007-500MG
101840-506EA 387.69 USD
101840-506
3-Methoxypyridin-2-ol
3-Methoxypyridin-2-ol

Hazard: Irritant

Formula: C₆H₇NO₂
MDL Number: MFCD08061315
CAS Number: 95907-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR