4-(Pyrrolidin-1-ylcarbonyl)pyridin-2-amine

Supplier: Matrix Scientific

036006-500MG
101840-504EA 230.77 USD
101840-504
4-(Pyrrolidin-1-ylcarbonyl)pyridin-2-amine
4-(Pyrrolidin-1-ylcarbonyl)pyridin-2-amine

Hazard: Irritant

Formula: C₁₀H₁₃N₃O
MDL Number: MFCD11053954
CAS Number: 1060817-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR