1-(2-Amino-5-methoxybenzyl)pyrrolidin-2-one

Supplier: Matrix Scientific

036005-500MG
101840-502EA 387.69 USD
101840-502
1-(2-Amino-5-methoxybenzyl)pyrrolidin-2-one
1-(2-Amino-5-methoxybenzyl)pyrrolidin-2-one

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD11696414
CAS Number: 1071351-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR