2-(Pyridin-2-ylthio)acetohydrazide

Supplier: Matrix Scientific

035999-500MG
101840-490EA 290.77 USD
101840-490
2-(Pyridin-2-ylthio)acetohydrazide
2-(Pyridin-2-ylthio)acetohydrazide

Hazard: Irritant

Formula: C₇H₉N₃OS
MDL Number: MFCD01422111
CAS Number: 104496-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR