N-(4-Bromophenyl)-4-hydrazino-4-oxobutanamide

Supplier: Matrix Scientific

035994-500MG
101840-480EA 217.35 USD
101840-480
N-(4-Bromophenyl)-4-hydrazino-4-oxobutanamide
N-(4-Bromophenyl)-4-hydrazino-4-oxobutanamide

Hazard: Irritant

Formula: C₁₀H₁₂BrN₃O₂
MDL Number: MFCD02255632
CAS Number: 443863-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR