2-[(4-Chlorophenyl)amino]acetohydrazide

Supplier: Matrix Scientific

035992-500MG
101840-476EA 87.4 USD
101840-476
2-[(4-Chlorophenyl)amino]acetohydrazide
2-[(4-Chlorophenyl)amino]acetohydrazide

Hazard: Irritant

Formula: C₈H₁₀ClN₃O
MDL Number: MFCD00463072
CAS Number: 2371-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR