2-(3-Methyl-1H-pyrazol-1-yl)acetohydrazide

Supplier: Matrix Scientific

035991-500MG
101840-474EA 289.8 USD
101840-474
2-(3-Methyl-1H-pyrazol-1-yl)acetohydrazide
2-(3-Methyl-1H-pyrazol-1-yl)acetohydrazide

Hazard: Irritant

Formula: C₆H₁₀N₄O
MDL Number: MFCD03419452
CAS Number: 512809-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR