3-(3,5-Dimethyl-1H-pyrazol-1-yl)propanohydrazide

Supplier: Matrix Scientific

035988-500MG
101840-468EA 217.35 USD
101840-468
3-(3,5-Dimethyl-1H-pyrazol-1-yl)propanohydrazide
3-(3,5-Dimethyl-1H-pyrazol-1-yl)propanohydrazide

Hazard: Irritant

Formula: C₈H₁₄N₄O
MDL Number: MFCD01954426
CAS Number: 313050-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR