2-[(3-Methylphenyl)amino]propanohydrazide

Supplier: Matrix Scientific

035987-500MG
101840-466EA 290.77 USD
101840-466
2-[(3-Methylphenyl)amino]propanohydrazide
2-[(3-Methylphenyl)amino]propanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₅N₃O
MDL Number: MFCD02255642
CAS Number: 1396996-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR