5-(1H-1,2,3-Benzotriazol-1-ylmethyl)-4-phenyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

035980-500MG
101840-452EA 290.77 USD
101840-452
5-(1H-1,2,3-Benzotriazol-1-ylmethyl)-4-phenyl-4H-1,2,4-triazole-3-thiol
5-(1H-1,2,3-Benzotriazol-1-ylmethyl)-4-phenyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₅H₁₂N₆S
MDL Number: MFCD01433963
CAS Number: 122503-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR