Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl 4-[(2,4-dichlorobenzyl)amino]benzoate

Supplier: Matrix Scientific


035976-500MG
101840-444EA 247.15 USD InStock
101840-444
Methyl 4-[(2,4-dichlorobenzyl)amino]benzoate
Methyl-4-[(2,4-dichlorobenzyl)amino]benzoate
CAS: 1071400-48-0
MDL Number: MFCD11696409
Molecular Formula: C15H13Cl2NO2
Molecular Weight: 310,18
Hazard: Irritant


Formula: C₁₅H₁₃Cl₂NO₂
MW: 310,18 g/mol
MDL Number: MFCD11696409
CAS Number: 1071400-48-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty