4-Fluoro-N-[4-(hydrazinocarbonyl)phenyl]benzamide

Supplier: Matrix Scientific

035970-500MG
101840-430EA 290.77 USD
101840-430
4-Fluoro-N-[4-(hydrazinocarbonyl)phenyl]benzamide
4-Fluoro-N-[4-(hydrazinocarbonyl)phenyl]benzamide

Hazard: Irritant

Formula: C₁₄H₁₂FN₃O₂
MDL Number: MFCD02255666
CAS Number: 172950-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR