2-[(3,4-Dimethylphenyl)amino]acetohydrazide

Supplier: Matrix Scientific

035967-500MG
101840-424EA 217.35 USD
101840-424
2-[(3,4-Dimethylphenyl)amino]acetohydrazide
2-[(3,4-Dimethylphenyl)amino]acetohydrazide

Hazard: Irritant

Formula: C₁₀H₁₅N₃O
MDL Number: MFCD03208903
CAS Number: 2370-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR