2-Chloro-N-[3-(hydrazinocarbonyl)phenyl]benzamide

Supplier: Matrix Scientific

035963-500MG
101840-416EA 290.77 USD
101840-416
2-Chloro-N-[3-(hydrazinocarbonyl)phenyl]benzamide
2-Chloro-N-[3-(hydrazinocarbonyl)phenyl]benzamide

Hazard: Irritant

Formula: C₁₄H₁₂ClN₃O₂
MDL Number: MFCD02255612
CAS Number: 1071296-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR