2-[(4-Methyl-5-phenyl-4H-1,2,4-triazol-3-yl)thio]acetohydrazide

Supplier: Matrix Scientific

035957-500MG
101840-406EA 290.77 USD
101840-406
2-[(4-Methyl-5-phenyl-4H-1,2,4-triazol-3-yl)thio]acetohydrazide
2-[(4-Methyl-5-phenyl-4H-1,2,4-triazol-3-yl)thio]acetohydrazide

Hazard: Irritant

Formula: C₁₁H₁₃N₅OS
MDL Number: MFCD02679122
CAS Number: 120873-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR