4-Chloro-N-[4-(hydrazinocarbonyl)phenyl]benzamide

Supplier: Matrix Scientific

035956-500MG
101840-404EA 290.77 USD
101840-404
4-Chloro-N-[4-(hydrazinocarbonyl)phenyl]benzamide
4-Chloro-N-[4-(hydrazinocarbonyl)phenyl]benzamide

Hazard: Irritant

Formula: C₁₄H₁₂ClN₃O₂
MDL Number: MFCD02255664
CAS Number: 100278-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR