4-Chloro-6-nitro-o-toluidine

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-methyl-6-nitroaniline

035952-500MG
101840-396EA 387.69 USD
101840-396
4-Chloro-6-nitro-o-toluidine
4-Chloro-6-nitro-o-toluidine

Hazard: Irritant

Formula: C₇H₇ClN₂O₂
MW: 186.60 g/mol
MDL Number: MFCD01320687
CAS Number: 62790-50-5
UN: 2237
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR