2-Amino-4-chloro-6-methylphenylamine

Supplier: Matrix Scientific

035951-500MG
101840-394EA 123.08 USD
101840-394
2-Amino-4-chloro-6-methylphenylamine
2-Amino-4-chloro-6-methylphenylamine

Hazard: Irritant

Formula: C₇H₉ClN₂
MDL Number: MFCD03426174
CAS Number: 109671-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR